missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dichlorobenzyl Alcohol 98.0+%, TCI America™
Supplier: TCI America D37915G
Specifications
| 3,5-Dichlorobenzyl Alcohol | |
| 79°C | |
| C7H6Cl2O | |
| 5 g | |
| VSNNLLQKDRCKCB-UHFFFAOYSA-N | |
| (3,5-dichlorophenyl)methanol | |
| 43236 | |
| ≥98.0% (GC) |
| 60211-57-6 | |
| White-Yellow | |
| MFCD00004634 | |
| 3,5-dichlorobenzyl alcohol, 3,5-dichlorophenyl methanol, benzenemethanol, 3,5-dichloro, 3,5-dichlorobenzylalcohol, 3,5-dichlorobenzylic alcohol, 3,5-dichlorobenzenemethanol, 3,5-dicholorbenzylalcohol, 3,5-dichlorophenyl methan-1-ol, 3, 5-dichlorobenzyl alcohol, pubchem4908 | |
| C1=C(C=C(C=C1Cl)Cl)CO | |
| 177.024 | |
| 177.02 | |
| Crystalline Powder |
Chemical Identifiers
| 60211-57-6 | |
| 177.024 | |
| VSNNLLQKDRCKCB-UHFFFAOYSA-N | |
| 43236 | |
| C1=C(C=C(C=C1Cl)Cl)CO |
| C7H6Cl2O | |
| MFCD00004634 | |
| 3,5-dichlorobenzyl alcohol, 3,5-dichlorophenyl methanol, benzenemethanol, 3,5-dichloro, 3,5-dichlorobenzylalcohol, 3,5-dichlorobenzylic alcohol, 3,5-dichlorobenzenemethanol, 3,5-dicholorbenzylalcohol, 3,5-dichlorophenyl methan-1-ol, 3, 5-dichlorobenzyl alcohol, pubchem4908 | |
| (3,5-dichlorophenyl)methanol |
Safety and Handling
TSCA : No