missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorobenzyl Alcohol 98.0+%, TCI America™

Catalog No. p-7127917
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37915G 5 g
D379125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D37915G Supplier TCI America Supplier No. D37915G
Only null left

Chemical Identifiers

CAS 60211-57-6
Molecular Formula C7H6Cl2O
Molecular Weight (g/mol) 177.024
MDL Number MFCD00004634
InChI Key VSNNLLQKDRCKCB-UHFFFAOYSA-N
Synonym 3,5-dichlorobenzyl alcohol, 3,5-dichlorophenyl methanol, benzenemethanol, 3,5-dichloro, 3,5-dichlorobenzylalcohol, 3,5-dichlorobenzylic alcohol, 3,5-dichlorobenzenemethanol, 3,5-dicholorbenzylalcohol, 3,5-dichlorophenyl methan-1-ol, 3, 5-dichlorobenzyl alcohol, pubchem4908
PubChem CID 43236
IUPAC Name (3,5-dichlorophenyl)methanol
SMILES C1=C(C=C(C=C1Cl)Cl)CO

Specifications

Melting Point 79°C
Color White-Yellow
Quantity 5 g
Formula Weight 177.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dichlorobenzyl Alcohol