missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorobenzoyl chloride, 96%

CAS: 2905-62-6 | C7H3Cl3O | 209.45 g/mol

$94.97 - $736.14

Chemical Identifiers

CAS 2905-62-6
Molecular Formula C7H3Cl3O
Molecular Weight (g/mol) 209.45
MDL Number MFCD00009817
InChI Key GGHLXLVPNZMBQR-UHFFFAOYSA-N
Synonym benzoyl chloride, 3,5-dichloro, 3,5-dichlorobenzoylchloride, dichlorobenzoylchloride 3,5-, pubchem10805, dsstox_cid_7506, acmc-209h6l, dsstox_rid_78478, dsstox_gsid_27506, ksc204k6p, 3,5-dichloro benzoyl chloride
PubChem CID 76191
IUPAC Name 3,5-dichlorobenzoyl chloride
SMILES ClC(=O)C1=CC(Cl)=CC(Cl)=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2467606
View Documents
Thermo Scientific Chemicals
B2467606
5 g
Each for $94.97
Only null left
 
AAB2467614
View Documents
Thermo Scientific Chemicals
B2467614
25 g
Each for $261.26
Only null left
 
AAB2467622
View Documents
Thermo Scientific Chemicals
B2467622
100 g
Each for $736.14
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2905-62-6
209.45
GGHLXLVPNZMBQR-UHFFFAOYSA-N
76191
ClC(=O)C1=CC(Cl)=CC(Cl)=C1
C7H3Cl3O
MFCD00009817
benzoyl chloride, 3,5-dichloro, 3,5-dichlorobenzoylchloride, dichlorobenzoylchloride 3,5-, pubchem10805, dsstox_cid_7506, acmc-209h6l, dsstox_rid_78478, dsstox_gsid_27506, ksc204k6p, 3,5-dichloro benzoyl chloride
3,5-dichlorobenzoyl chloride

Specifications

2905-62-6
1.478
137°C (278°F)
1.582
5 g
1940681
benzoyl chloride, 3,5-dichloro, 3,5-dichlorobenzoylchloride, dichlorobenzoylchloride 3,5-, pubchem10805, dsstox_cid_7506, acmc-209h6l, dsstox_rid_78478, dsstox_gsid_27506, ksc204k6p, 3,5-dichloro benzoyl chloride
ClC(=O)C1=CC(Cl)=CC(Cl)=C1
209.45
209.46
3,5-Dichlorobenzoyl chloride
24°C to 28°C
245°C
C7H3Cl3O
MFCD00009817
UN3261
Moisture sensitive
GGHLXLVPNZMBQR-UHFFFAOYSA-N
3,5-dichlorobenzoyl chloride
76191
96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 220-813-6

RTECSNumber : DM6636766

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only