Learn More
3,5-Dichlorobenzoyl chloride, 96%
CAS: 2905-62-6 | C7H3Cl3O | 209.45 g/mol
Supplier: Thermo Scientific Chemicals B2467614
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 3,5-Dichlorobenzoyl chloride | |
| 24°C to 28°C | |
| 245°C | |
| C7H3Cl3O | |
| MFCD00009817 | |
| UN3261 | |
| Moisture sensitive | |
| GGHLXLVPNZMBQR-UHFFFAOYSA-N | |
| 3,5-dichlorobenzoyl chloride | |
| 76191 | |
| 96% |
| 2905-62-6 | |
| 1.478 | |
| 137°C (278°F) | |
| 1.582 | |
| 25 g | |
| 1940681 | |
| benzoyl chloride, 3,5-dichloro, 3,5-dichlorobenzoylchloride, dichlorobenzoylchloride 3,5-, pubchem10805, dsstox_cid_7506, acmc-209h6l, dsstox_rid_78478, dsstox_gsid_27506, ksc204k6p, 3,5-dichloro benzoyl chloride | |
| ClC(=O)C1=CC(Cl)=CC(Cl)=C1 | |
| 209.45 | |
| 209.46 |
Chemical Identifiers
| 2905-62-6 | |
| 209.45 | |
| GGHLXLVPNZMBQR-UHFFFAOYSA-N | |
| 76191 | |
| ClC(=O)C1=CC(Cl)=CC(Cl)=C1 |
| C7H3Cl3O | |
| MFCD00009817 | |
| benzoyl chloride, 3,5-dichloro, 3,5-dichlorobenzoylchloride, dichlorobenzoylchloride 3,5-, pubchem10805, dsstox_cid_7506, acmc-209h6l, dsstox_rid_78478, dsstox_gsid_27506, ksc204k6p, 3,5-dichloro benzoyl chloride | |
| 3,5-dichlorobenzoyl chloride |
Safety and Handling
GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 220-813-6
RTECSNumber : DM6636766
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only