missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromotoluene 98.0+%, TCI America™

Catalog No. p-7127701
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33765G 5 g
D337625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33765G Supplier TCI America Supplier No. D33765G

Chemical Identifiers

CAS 1611-92-3
Molecular Formula C7H6Br2
Molecular Weight (g/mol) 249.93
MDL Number MFCD00013528
InChI Key DPKKOVGCHDUSAI-UHFFFAOYSA-N
Synonym 3,5-dibromotoluene, benzene, 1,3-dibromo-5-methyl, toluene, 3,5-dibromo, 3 ,5-dibromotoluene, 1,3-dibromo-5-methyl-benzene, 5-dibromotoluene, 3,5-dibromo toluene, 3,5-dibromo-toluene, pubchem3934, acmc-1bs6w
PubChem CID 15361
IUPAC Name 1,3-dibromo-5-methylbenzene
SMILES CC1=CC(Br)=CC(Br)=C1

Specifications

Melting Point 41°C
Color White-Yellow
Boiling Point 246°C
Quantity 5 g
Formula Weight 249.93
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dibromotoluene