accessibility menu, dialog, popup

UserName

CH3BrBr

CAS RN 1611-92-3

IUPAC Name: 1,3-dibromo-5-methylbenzene
Synonyms: 1,3-dibromo-5-methylbenzene
Molecular Weight (g/mol): 249.93
Molecular Formula: C7H6Br2
InChi Key: DPKKOVGCHDUSAI-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=CC(Br)=C1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (5)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)