missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromotoluene, 98+%

Catalog No. p-7021430
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1375606 5 g
AAA1375614 25 g
AAA1375622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1375606 Supplier Thermo Scientific Chemicals Supplier No. A1375606

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1611-92-3 | C7H6Br2 | 249.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1611-92-3
Molecular Formula C7H6Br2
Molecular Weight (g/mol) 249.93
MDL Number MFCD00013528
InChI Key DPKKOVGCHDUSAI-UHFFFAOYSA-N
Synonym 3,5-dibromotoluene, benzene, 1,3-dibromo-5-methyl, toluene, 3,5-dibromo, 3 ,5-dibromotoluene, 1,3-dibromo-5-methyl-benzene, 5-dibromotoluene, 3,5-dibromo toluene, 3,5-dibromo-toluene, pubchem3934, acmc-1bs6w
PubChem CID 15361
IUPAC Name 1,3-dibromo-5-methylbenzene
SMILES CC1=CC(Br)=CC(Br)=C1

Specifications

Melting Point 37°C to 41°C
Boiling Point 246°C
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 1928685
Formula Weight 249.94
Percent Purity ≥98%
Chemical Name or Material 3,5-Dibromotoluene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only