missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dibromopyridine 98.0+%, TCI America™
$69.31 - $217.54
Chemical Identifiers
| CAS | 625-92-3 |
|---|---|
| Molecular Formula | C5H3Br2N |
| Molecular Weight (g/mol) | 236.89 |
| MDL Number | MFCD00014634 |
| InChI Key | SOSPMXMEOFGPIM-UHFFFAOYSA-N |
| Synonym | pyridine, 3,5-dibromo, 3,5-dibromo-pyridine, 3,5-dibromo pyridine, 3,5-dibrompyridin, zlchem 300, 3.5-dibromopyridine, 3,5-dibromo-pyridin, pubchem2133, 3, 5-dibromopyridine, acmc-1auln |
| PubChem CID | 69369 |
| ChEBI | CHEBI:51593 |
| IUPAC Name | 3,5-dibromopyridine |
| SMILES | BrC1=CC(Br)=CN=C1 |
Chemical Identifiers
| 625-92-3 | |
| 236.89 | |
| SOSPMXMEOFGPIM-UHFFFAOYSA-N | |
| 69369 | |
| 3,5-dibromopyridine |
| C5H3Br2N | |
| MFCD00014634 | |
| pyridine, 3,5-dibromo, 3,5-dibromo-pyridine, 3,5-dibromo pyridine, 3,5-dibrompyridin, zlchem 300, 3.5-dibromopyridine, 3,5-dibromo-pyridin, pubchem2133, 3, 5-dibromopyridine, acmc-1auln | |
| CHEBI:51593 | |
| BrC1=CC(Br)=CN=C1 |
Specifications
| 625-92-3 | |
| White-Yellow | |
| C5H3Br2N | |
| 5 g | |
| SOSPMXMEOFGPIM-UHFFFAOYSA-N | |
| 3,5-dibromopyridine | |
| 69369 | |
| 236.89 | |
| Crystalline Powder |
| 111°C | |
| 222°C | |
| MFCD00014634 | |
| pyridine, 3,5-dibromo, 3,5-dibromo-pyridine, 3,5-dibromo pyridine, 3,5-dibrompyridin, zlchem 300, 3.5-dibromopyridine, 3,5-dibromo-pyridin, pubchem2133, 3, 5-dibromopyridine, acmc-1auln | |
| BrC1=CC(Br)=CN=C1 | |
| 236.89 | |
| CHEBI:51593 | |
| ≥98.0% (GC) | |
| 3,5-Dibromopyridine |
Safety and Handling
TSCA : No