missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dibromopyridine 98.0+%, TCI America™
Supplier: TCI America D257325G
Specifications
| 3,5-Dibromopyridine | |
| 111°C | |
| 222°C | |
| MFCD00014634 | |
| pyridine, 3,5-dibromo, 3,5-dibromo-pyridine, 3,5-dibromo pyridine, 3,5-dibrompyridin, zlchem 300, 3.5-dibromopyridine, 3,5-dibromo-pyridin, pubchem2133, 3, 5-dibromopyridine, acmc-1auln | |
| BrC1=CC(Br)=CN=C1 | |
| 236.89 | |
| CHEBI:51593 | |
| ≥98.0% (GC) |
| 625-92-3 | |
| White-Yellow | |
| C5H3Br2N | |
| 25 g | |
| SOSPMXMEOFGPIM-UHFFFAOYSA-N | |
| 3,5-dibromopyridine | |
| 69369 | |
| 236.89 | |
| Crystalline Powder |
Chemical Identifiers
| 625-92-3 | |
| 236.89 | |
| SOSPMXMEOFGPIM-UHFFFAOYSA-N | |
| 69369 | |
| 3,5-dibromopyridine |
| C5H3Br2N | |
| MFCD00014634 | |
| pyridine, 3,5-dibromo, 3,5-dibromo-pyridine, 3,5-dibromo pyridine, 3,5-dibrompyridin, zlchem 300, 3.5-dibromopyridine, 3,5-dibromo-pyridin, pubchem2133, 3, 5-dibromopyridine, acmc-1auln | |
| CHEBI:51593 | |
| BrC1=CC(Br)=CN=C1 |
Safety and Handling
TSCA : No