accessibility menu, dialog, popup

UserName

CAS RN 625-92-3

BrNBr

CAS RN 625-92-3

IUPAC Name: 3,5-dibromopyridine
Synonyms: 3,5-dibromopyridine
Molecular Weight (g/mol): 236.89
Molecular Formula: C5H3Br2N
InChi Key: SOSPMXMEOFGPIM-UHFFFAOYSA-N
SMILES: BrC1=CC(Br)=CN=C1
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (3)
  • (2)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (6)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)