missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromobenzotrifluoride 98.0+%, TCI America™

Catalog No. D480725G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D480725G 25 g
D48075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D480725G Supplier TCI America Supplier No. D480725G

Chemical Identifiers

CAS 401-84-3
Molecular Formula C7H3Br2F3
Molecular Weight (g/mol) 303.90
MDL Number MFCD00526219
InChI Key OFNXSUANJLHGQN-UHFFFAOYSA-N
Synonym 1,3-dibromo-5-trifluoromethyl benzene, 3,5-dibromobenzotrifluoride, benzene, 1,3-dibromo-5-trifluoromethyl, pubchem11234, acmc-209jbn, ksc911m2p, 1,3-dibromobenzotrifluoride, 3,5-dibromo-, ,-trifluorotoluene, 3,5-dibromo-1-trifluoromethyl benzene, 1,3-dibromo-5-trifluoromethyl ;benzene
PubChem CID 2063489
IUPAC Name 1,3-dibromo-5-(trifluoromethyl)benzene
SMILES FC(F)(F)C1=CC(Br)=CC(Br)=C1

Specifications

Color Yellow
Boiling Point 211°C
Quantity 25 g
Formula Weight 303.90
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,5-Dibromobenzotrifluoride