missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromobenzoic Acid 97.0+%, TCI America™

Catalog No. p-7127250
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23535G 5 g
D235310G 10 g
D235325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D23535G Supplier TCI America Supplier No. D23535G

Chemical Identifiers

CAS 618-58-6
Molecular Formula C7H4Br2O2
Molecular Weight (g/mol) 279.92
MDL Number MFCD00051758
InChI Key SFTFNJZWZHASAQ-UHFFFAOYSA-N
Synonym benzoic acid, 3,5-dibromo, 3,5-dibromobenzoicacid, benzoicacid, 3,5-dibromo, buttpark 99\57-19, pubchem3861, acmc-209mwz, maybridge1_003318, 3,5-dibromo-benzoic acid, 4-09-00-01028 beilstein handbook reference, ksc358a3h
PubChem CID 12063
IUPAC Name 3,5-dibromobenzoic acid
SMILES OC(=O)C1=CC(Br)=CC(Br)=C1

Specifications

Melting Point 220°C
Color White-Yellow
Quantity 5 g
Formula Weight 279.92
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dibromobenzoic Acid
EINECSNumber (3)-0062
RTECSNumber DG6290010
TSCA No