accessibility menu, dialog, popup

UserName

CAS RN 618-58-6

IUPAC Name: 3,5-dibromobenzoic acid
Synonyms: 3,5-dibromobenzoic acid
Molecular Weight (g/mol): 279.92
Molecular Formula: C7H4Br2O2
InChi Key: SFTFNJZWZHASAQ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(Br)=CC(Br)=C1
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results