missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromobenzeneboronic acid, 97%

Catalog No. AAB2386306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2386306 5 g
AAB2386314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2386306 Supplier Thermo Scientific Chemicals Supplier No. B2386306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 117695-55-3 | C6H5BBr2O2 | 279.722 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 117695-55-3
Molecular Formula C6H5BBr2O2
Molecular Weight (g/mol) 279.722
MDL Number MFCD01075725
InChI Key WQBLCGDZYFKINX-UHFFFAOYSA-N
Synonym 3,5-dibromobenzeneboronic acid, 3,5-dibromophenyl boronic acid, boronic acid, 3,5-dibromophenyl, pubchem14108, acmc-2099ur, amtb596, 3,5-dibromobenzeneboronicacid, ksc493i3d, 3,5-dibromobenzene boronic acid
PubChem CID 2734689
IUPAC Name (3,5-dibromophenyl)boronic acid
SMILES B(C1=CC(=CC(=C1)Br)Br)(O)O

Specifications

Melting Point >300°C
Quantity 5 g
Beilstein 6645285
Formula Weight 279.73
Percent Purity 97%
Chemical Name or Material 3,5-Dibromobenzeneboronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only