missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibenzyloxybenzoic Acid 98.0+%, TCI America™

Catalog No. D22675G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22675G 5 g
D226725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D22675G Supplier TCI America Supplier No. D22675G
Only null left

Chemical Identifiers

CAS 28917-43-3
Molecular Formula C21H18O4
Molecular Weight (g/mol) 334.371
MDL Number MFCD00191341
InChI Key DHQIBPUGSWVDOH-UHFFFAOYSA-N
Synonym 3,5-Bis(benzyloxy)benzoic Acid
PubChem CID 99307
IUPAC Name 3,5-bis(phenylmethoxy)benzoic acid
SMILES C1=CC=C(C=C1)COC2=CC(=CC(=C2)C(=O)O)OCC3=CC=CC=C3

Specifications

Melting Point 214°C
Color White-Yellow
Quantity 5 g
Formula Weight 334.37
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dibenzyloxybenzoic Acid