CAS RN 28917-43-3
IUPAC Name:
3,5-bis(benzyloxy)benzoic acid
Synonyms:
3,5-bis(benzyloxy)benzoic acid
Molecular Weight (g/mol):
334.37
Molecular Formula:
C21H18O4
InChi Key:
DHQIBPUGSWVDOH-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC(OCC2=CC=CC=C2)=CC(OCC2=CC=CC=C2)=C1
1
–
1
of
1
results
1
3,5-Dibenzyloxybenzoic Acid 98.0+%, TCI America™
CAS: 28917-43-3 Molecular Formula: C21H18O4 Molecular Weight (g/mol): 334.371 MDL Number: MFCD00191341 InChI Key: DHQIBPUGSWVDOH-UHFFFAOYSA-N Synonym: 3,5-Bis(benzyloxy)benzoic Acid PubChem CID: 99307 IUPAC Name: 3,5-bis(phenylmethoxy)benzoic acid SMILES: C1=CC=C(C=C1)COC2=CC(=CC(=C2)C(=O)O)OCC3=CC=CC=C3