Learn More
3,5-Diaminobenzoic acid, 96%, may contain up to 3% moisture
CAS: 535-87-5 | C7H8N2O2 | 152.153 g/mol
Supplier: Thermo Scientific Chemicals A1407522
Description
3,5-Diaminobenzoic acid is an analog of diatriazoate, which has been shown to have antigenicity properties towards the monoclonal antibodies. It is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,5-Diaminobenzoic acid is an analog of diatriazoate, which has been shown to have antigenicity properties towards the monoclonal antibodies. It is used as pharmaceutical intermediate.
Solubility
Very soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
| 3,5-Diaminobenzoic acid | |
| ∼235°C (decomposition) | |
| may contain up to 3% moisture | |
| MFCD00007807 | |
| 2086484 | |
| benzoic acid, 3,5-diamino, 1-carboxy-3,5-diaminobenzene, ccris 2885, 3,5-diamino benzoic acid, 3,5-diaminobenzoicacid, dabz, pubchem21025, 3,5-diaminebenzoicacid, acmc-1awxi, 3,5-diamino-benzoic acid | |
| UENRXLSRMCSUSN-UHFFFAOYSA-N | |
| 3,5-diaminobenzoic acid | |
| 12062 | |
| 98% |
| 535-87-5 | |
| 210°C (410°F) | |
| C7H8N2O2 | |
| 100 g | |
| 14,2978 | |
| Very soluble in water. | |
| C1=C(C=C(C=C1N)N)C(=O)O | |
| 152.153 | |
| 152.15 |
Chemical Identifiers
| 535-87-5 | |
| 152.153 | |
| UENRXLSRMCSUSN-UHFFFAOYSA-N | |
| 12062 | |
| C1=C(C=C(C=C1N)N)C(=O)O |
| C7H8N2O2 | |
| MFCD00007807 | |
| benzoic acid, 3,5-diamino, 1-carboxy-3,5-diaminobenzene, ccris 2885, 3,5-diamino benzoic acid, 3,5-diaminobenzoicacid, dabz, pubchem21025, 3,5-diaminebenzoicacid, acmc-1awxi, 3,5-diamino-benzoic acid | |
| 3,5-diaminobenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 208-621-0
RTECSNumber : DG6186000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only