accessibility menu, dialog, popup

UserName

CAS RN 535-87-5

IUPAC Name: 5-carboxybenzene-1,3-bis(aminium) dichloride
Synonyms: 5-carboxybenzene-1,3-bis(aminium) dichloride
Molecular Weight (g/mol): 225.07
Molecular Formula: C7H10Cl2N2O2
InChi Key: IGPLRBRBTUNCRT-UHFFFAOYSA-N
SMILES: [Cl-].[Cl-].[NH3+]C1=CC(=CC([NH3+])=C1)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (2)
  • (4)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)