missing translation for 'onlineSavingsMsg'
Learn More

3,5-Diaminobenzoic acid, 96%, may contain up to 3% moisture

Catalog No. AAA1407536
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1407536 500 g
AAA1407518 50 g
AAA1407522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1407536 Supplier Thermo Scientific Chemicals Supplier No. A1407536
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 535-87-5 | C7H8N2O2 | 152.153 g/mol

3,5-Diaminobenzoic acid is an analog of diatriazoate, which has been shown to have antigenicity properties towards the monoclonal antibodies. It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Diaminobenzoic acid is an analog of diatriazoate, which has been shown to have antigenicity properties towards the monoclonal antibodies. It is used as pharmaceutical intermediate.

Solubility
Very soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 535-87-5
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.153
MDL Number MFCD00007807
InChI Key UENRXLSRMCSUSN-UHFFFAOYSA-N
Synonym benzoic acid, 3,5-diamino, 1-carboxy-3,5-diaminobenzene, ccris 2885, 3,5-diamino benzoic acid, 3,5-diaminobenzoicacid, dabz, pubchem21025, 3,5-diaminebenzoicacid, acmc-1awxi, 3,5-diamino-benzoic acid
PubChem CID 12062
IUPAC Name 3,5-diaminobenzoic acid
SMILES C1=C(C=C(C=C1N)N)C(=O)O

Specifications

Melting Point ∼235°C (decomposition)
Flash Point 210°C (410°F)
Quantity 500 g
Beilstein 2086484
Merck Index 14,2978
Solubility Information Very soluble in water.
Formula Weight 152.15
Percent Purity 98%
Assay may contain up to 3% moisture
Chemical Name or Material 3,5-Diaminobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 208-621-0
RTECSNumber DG6186000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only