missing translation for 'onlineSavingsMsg'
Learn More

3,5-Diacetoxybenzoic Acid 98.0+%, TCI America™

Catalog No. D28441G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28441G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D28441G Supplier TCI America Supplier No. D28441G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 35354-29-1
Molecular Formula C11H10O6
Molecular Weight (g/mol) 238.195
MDL Number MFCD00017591
InChI Key QBTDQJMLMVEUTQ-UHFFFAOYSA-N
Synonym 3,5-diacetoxybenzoic acid, 3,5-bis acetyloxy benzoic acid, 3,5-diacethoxybenzoic acid, benzoic acid, 3,5-bis acetyloxy, 3,5-diacetoxy benzoic acid, maybridge1_004330, acmc-209igc, 3.5-diacetoxybenzoic acid, 3,5-diacetyloxybezoic acid, 3,5-diacetoxy-benzoic acid
PubChem CID 539869
IUPAC Name 3,5-diacetyloxybenzoic acid
SMILES CC(=O)OC1=CC(=CC(=C1)C(=O)O)OC(=O)C

Specifications

Melting Point 161°C
Color White-Yellow
Quantity 1 g
Formula Weight 238.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Diacetoxybenzoic Acid