accessibility menu, dialog, popup

UserName

CAS RN 35354-29-1

IUPAC Name: 3,5-bis(acetyloxy)benzoic acid
Synonyms: 3,5-bis(acetyloxy)benzoic acid
Molecular Weight (g/mol): 238.2
Molecular Formula: C11H10O6
InChi Key: QBTDQJMLMVEUTQ-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC(=CC(OC(C)=O)=C1)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results