missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-tert-butylsalicylic Acid 95.0+%, TCI America™

Catalog No. D1947500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1947500G 500 g
D194725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1947500G Supplier TCI America Supplier No. D1947500G

Chemical Identifiers

CAS 19715-19-6
Molecular Formula C15H22O3
Molecular Weight (g/mol) 250.338
MDL Number MFCD00059058
InChI Key ZWQBZEFLFSFEOS-UHFFFAOYSA-N
Synonym 3,5-di-tert-butyl-2-hydroxybenzoic acid, 3,5-bis-tert-butylsalicylic acid, 3,5-di-tert-butylsalicylic acid, benzoic acid, 3,5-bis 1,1-dimethylethyl hydroxy, benzoic acid, 3,5-bis 1,1-dimethylethyl-2-hydroxy, 3,5-ditertbutyl salicylic acid, dibutylsalicylicacid, acmc-1c7qv, 3,2-bis 1,1-dimethylethyl-2-hydroxybenzoic acid, ksc466o9n
PubChem CID 88208
IUPAC Name 3,5-ditert-butyl-2-hydroxybenzoic acid
SMILES CC(C)(C)C1=CC(=C(C(=C1)C(=O)O)O)C(C)(C)C

Specifications

Melting Point 163°C
Color White-Yellow
Quantity 500 g
Formula Weight 250.34
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Di-tert-butylsalicylic Acid
EINECSNumber (3)-3034
TSCA Yes