accessibility menu, dialog, popup

UserName

CH3H3CCH3OHOOHH3CH3CCH3

CAS RN 19715-19-6

IUPAC Name: 3,5-di-tert-butyl-2-hydroxybenzoic acid
Synonyms: 3,5-di-tert-butyl-2-hydroxybenzoic acid
Molecular Weight (g/mol): 250.34
Molecular Formula: C15H22O3
InChi Key: ZWQBZEFLFSFEOS-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(C(O)=O)=C(O)C(=C1)C(C)(C)C
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)