missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-tert-butylcatechol, 99%

CAS: 1020-31-1 | C14H22O2 | 222.328 g/mol

$67.96 - $224.19

Chemical Identifiers

CAS 1020-31-1
Molecular Formula C14H22O2
Molecular Weight (g/mol) 222.328
MDL Number MFCD00008819
InChI Key PJZLSMMERMMQBJ-UHFFFAOYSA-N
Synonym 3,5-di-tert-butylcatechol, 3,5-di-t-butylcatechol, 3,5-di-tert-butylbenzene-1,2-diol, 3,5-di-tert-butylpyrocatechol, 1,2-benzenediol, 3,5-bis 1,1-dimethylethyl, 4,6-di-tert-butylpyrocatechol, unii-1328kn3f8b, 1,2-benzenediol, 3,5-di 1,1-dimethylethyl, pyrocatechol, 3,5-di-tert-butyl, 3,5-bis tert-butyl benzene-1,2-diol
PubChem CID 66099
IUPAC Name 3,5-ditert-butylbenzene-1,2-diol
SMILES CC(C)(C)C1=CC(=C(C(=C1)O)O)C(C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5520706
View Documents
Thermo Scientific Chemicals
H5520706
5 g
Each for $67.96
Only null left
 
AAH5520714
View Documents
Thermo Scientific Chemicals
H5520714
25 g
Each for $224.19
Only null left
 
Description

Description

It is an important raw material and intermediate used in organic synthesis, Pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, Pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Insoluble in water.

Notes
Store at room temperature with proper ventilation. Keep away from oxidizing agents.
Specifications

Chemical Identifiers

1020-31-1
222.328
PJZLSMMERMMQBJ-UHFFFAOYSA-N
66099
CC(C)(C)C1=CC(=C(C(=C1)O)O)C(C)(C)C
C14H22O2
MFCD00008819
3,5-di-tert-butylcatechol, 3,5-di-t-butylcatechol, 3,5-di-tert-butylbenzene-1,2-diol, 3,5-di-tert-butylpyrocatechol, 1,2-benzenediol, 3,5-bis 1,1-dimethylethyl, 4,6-di-tert-butylpyrocatechol, unii-1328kn3f8b, 1,2-benzenediol, 3,5-di 1,1-dimethylethyl, pyrocatechol, 3,5-di-tert-butyl, 3,5-bis tert-butyl benzene-1,2-diol
3,5-ditert-butylbenzene-1,2-diol

Specifications

1020-31-1
Brown to White
MFCD00008819
1370212
Insoluble in water.
CC(C)(C)C1=CC(=C(C(=C1)O)O)C(C)(C)C
222.328
222.33
Solid
95°C to 100°C
C14H22O2
5 g
3,5-di-tert-butylcatechol, 3,5-di-t-butylcatechol, 3,5-di-tert-butylbenzene-1,2-diol, 3,5-di-tert-butylpyrocatechol, 1,2-benzenediol, 3,5-bis 1,1-dimethylethyl, 4,6-di-tert-butylpyrocatechol, unii-1328kn3f8b, 1,2-benzenediol, 3,5-di 1,1-dimethylethyl, pyrocatechol, 3,5-di-tert-butyl, 3,5-bis tert-butyl benzene-1,2-diol
PJZLSMMERMMQBJ-UHFFFAOYSA-N
3,5-ditert-butylbenzene-1,2-diol
66099
99%
3,5-Di-tert-butylcatechol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 213-816-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only