accessibility menu, dialog, popup

UserName

CAS RN 1020-31-1

IUPAC Name: 3,5-di-tert-butylbenzene-1,2-diol
Synonyms: 3,5-di-tert-butylbenzene-1,2-diol
Molecular Weight (g/mol): 222.33
Molecular Formula: C14H22O2
InChi Key: PJZLSMMERMMQBJ-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(O)=C(O)C(=C1)C(C)(C)C
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (3)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (4)
  • (3)
  • (2)
  • (1)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)