missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-tert-butylbenzyl Bromide 98.0+%, TCI America™

Catalog No. D26535G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26535G 5 g
D26531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D26535G Supplier TCI America Supplier No. D26535G
Only null left

Chemical Identifiers

CAS 62938-08-3
Molecular Formula C15H23Br
Molecular Weight (g/mol) 283.25
MDL Number MFCD03701623
InChI Key SNRYBGHMHAJTTM-UHFFFAOYSA-N
Synonym alpha-Bromo-3,5-di-tert-butyltoluene
PubChem CID 4447507
IUPAC Name 1-(bromomethyl)-3,5-di-tert-butylbenzene
SMILES CC(C)(C)C1=CC(=CC(CBr)=C1)C(C)(C)C

Specifications

Color White
Quantity 5 g
UN Number 1759
Formula Weight 283.25
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material 3,5-Di-tert-butylbenzyl Bromide