CAS RN 62938-08-3
IUPAC Name:
1-(bromomethyl)-3,5-di-tert-butylbenzene
Synonyms:
1-(bromomethyl)-3,5-di-tert-butylbenzene
Molecular Weight (g/mol):
283.25
Molecular Formula:
C15H23Br
InChi Key:
SNRYBGHMHAJTTM-UHFFFAOYSA-N
SMILES:
CC(C)(C)C1=CC(=CC(CBr)=C1)C(C)(C)C
1
–
1
of
1
results
1
3,5-Di-tert-butylbenzyl Bromide 98.0+%, TCI America™
CAS: 62938-08-3 Molecular Formula: C15H23Br Molecular Weight (g/mol): 283.25 MDL Number: MFCD03701623 InChI Key: SNRYBGHMHAJTTM-UHFFFAOYSA-N Synonym: alpha-Bromo-3,5-di-tert-butyltoluene PubChem CID: 4447507 IUPAC Name: 1-(bromomethyl)-3,5-di-tert-butylbenzene SMILES: CC(C)(C)C1=CC(=CC(CBr)=C1)C(C)(C)C