missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di(tert-butyl)-4-hydroxybenzonitrile, 97%, Thermo Scientific™

Catalog No. p-2699439
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BTB09888DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTB09888DA Supplier Thermo Scientific Chemicals Supplier No. BTB09888DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1988-88-1
Molecular Formula C15H21NO
Molecular Weight (g/mol) 231.34
MDL Number MFCD00156137
InChI Key AKXIIOLURNATOC-UHFFFAOYSA-N
Synonym 3,5-di-tert-butyl-4-hydroxybenzonitrile, 3,5-di tert-butyl-4-hydroxybenzonitrile, benzonitrile, 3,5-bis 1,1-dimethylethyl-4-hydroxy, benzonitrile, 3,5-di-tert-butyl-4-hydroxy, 4-cyano-2,6-di-tert-butylphenol, 3,5-bis tert-butyl-4-hydroxybenzenecarbonitrile, acmc-20a6ry, 2,6-bis tert-butyl-4-cyanophenol, 3,5-di-t-butyl-4-hydroxybenzonitrile, 4-hydroxy-3,5-di-t-butylbenzonitrile
PubChem CID 137274
SMILES CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)C#N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
IUPAC Name 3,5-ditert-butyl-4-hydroxybenzonitrile
Formula Weight 231.34
Percent Purity 97%
Chemical Name or Material 3, 5-Di(tert-butyl)-4-hydroxybenzonitrile
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.