CAS RN 1988-88-1
IUPAC Name:
3,5-di-tert-butyl-4-hydroxybenzonitrile
Synonyms:
3,5-di-tert-butyl-4-hydroxybenzonitrile
Molecular Weight (g/mol):
231.34
Molecular Formula:
C15H21NO
InChi Key:
AKXIIOLURNATOC-UHFFFAOYSA-N
SMILES:
CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)C#N
1
–
3
of
3
results
1
3,5-Di(tert-butyl)-4-hydroxybenzonitrile, 97%, Thermo Scientific™
CAS: 1988-88-1 Molecular Formula: C15H21NO Molecular Weight (g/mol): 231.34 MDL Number: MFCD00156137 InChI Key: AKXIIOLURNATOC-UHFFFAOYSA-N Synonym: 3,5-di-tert-butyl-4-hydroxybenzonitrile,3,5-di tert-butyl-4-hydroxybenzonitrile,benzonitrile, 3,5-bis 1,1-dimethylethyl-4-hydroxy,benzonitrile, 3,5-di-tert-butyl-4-hydroxy,4-cyano-2,6-di-tert-butylphenol,3,5-bis tert-butyl-4-hydroxybenzenecarbonitrile,acmc-20a6ry,2,6-bis tert-butyl-4-cyanophenol,3,5-di-t-butyl-4-hydroxybenzonitrile,4-hydroxy-3,5-di-t-butylbenzonitrile PubChem CID: 137274 SMILES: CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)C#N