missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Di-tert-butyl-1,2-benzoquinone 98.0+%, TCI America™
$172.77 - $510.72
Chemical Identifiers
| CAS | 3383-21-9 |
|---|---|
| Molecular Formula | C14H20O2 |
| Molecular Weight (g/mol) | 220.312 |
| MDL Number | MFCD00001647 |
| InChI Key | NOUZOVBGCDDMSX-UHFFFAOYSA-N |
| Synonym | 3,5-di-tert-butyl-o-benzoquinone, 3,5-di-tert-butyl-1,2-benzoquinone, 3,5-di tert-butyl benzo-1,2-quinone, 3,5-cyclohexadiene-1,2-dione, 3,5-bis 1,1-dimethylethyl, 3,5-di-tert-butylcyclohexa-3,5-diene-1,2-dione, o-benzoquinone, 3,5-di-tert-butyl, 3,5-di-tert-butylbenzo-1,2-quinone, 3,5-di-t-butyl-1,2-benzoquinone, 3,5-ditert-butylbenzo-1,2-quinone, 3,5-bis tert-butyl cyclohexa-3,5-diene-1,2-dione |
| PubChem CID | 76915 |
| IUPAC Name | 3,5-ditert-butylcyclohexa-3,5-diene-1,2-dione |
| SMILES | CC(C)(C)C1=CC(=O)C(=O)C(=C1)C(C)(C)C |
Chemical Identifiers
| C14H20O2 | |
| MFCD00001647 | |
| 3,5-di-tert-butyl-o-benzoquinone, 3,5-di-tert-butyl-1,2-benzoquinone, 3,5-di tert-butyl benzo-1,2-quinone, 3,5-cyclohexadiene-1,2-dione, 3,5-bis 1,1-dimethylethyl, 3,5-di-tert-butylcyclohexa-3,5-diene-1,2-dione, o-benzoquinone, 3,5-di-tert-butyl, 3,5-di-tert-butylbenzo-1,2-quinone, 3,5-di-t-butyl-1,2-benzoquinone, 3,5-ditert-butylbenzo-1,2-quinone, 3,5-bis tert-butyl cyclohexa-3,5-diene-1,2-dione | |
| 3,5-ditert-butylcyclohexa-3,5-diene-1,2-dione |
Specifications
| 3383-21-9 | |
| C14H20O2 | |
| 3,5-di-tert-butyl-o-benzoquinone, 3,5-di-tert-butyl-1,2-benzoquinone, 3,5-di tert-butyl benzo-1,2-quinone, 3,5-cyclohexadiene-1,2-dione, 3,5-bis 1,1-dimethylethyl, 3,5-di-tert-butylcyclohexa-3,5-diene-1,2-dione, o-benzoquinone, 3,5-di-tert-butyl, 3,5-di-tert-butylbenzo-1,2-quinone, 3,5-di-t-butyl-1,2-benzoquinone, 3,5-ditert-butylbenzo-1,2-quinone, 3,5-bis tert-butyl cyclohexa-3,5-diene-1,2-dione | |
| CC(C)(C)C1=CC(=O)C(=O)C(=C1)C(C)(C)C | |
| 220.312 | |
| 220.31 | |
| Crystalline Powder |
| 115°C | |
| MFCD00001647 | |
| NOUZOVBGCDDMSX-UHFFFAOYSA-N | |
| 3,5-ditert-butylcyclohexa-3,5-diene-1,2-dione | |
| 76915 | |
| ≥98.0% (GC) | |
| 3,5-Di-tert-butyl-1,2-benzoquinone |
Safety and Handling
TSCA : No