missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-tert-butyl-1,2-benzoquinone 98.0+%, TCI America™

Catalog No. D24305G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24305G 5 g
D243025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D24305G Supplier TCI America Supplier No. D24305G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3383-21-9
Molecular Formula C14H20O2
Molecular Weight (g/mol) 220.312
MDL Number MFCD00001647
InChI Key NOUZOVBGCDDMSX-UHFFFAOYSA-N
Synonym 3,5-di-tert-butyl-o-benzoquinone, 3,5-di-tert-butyl-1,2-benzoquinone, 3,5-di tert-butyl benzo-1,2-quinone, 3,5-cyclohexadiene-1,2-dione, 3,5-bis 1,1-dimethylethyl, 3,5-di-tert-butylcyclohexa-3,5-diene-1,2-dione, o-benzoquinone, 3,5-di-tert-butyl, 3,5-di-tert-butylbenzo-1,2-quinone, 3,5-di-t-butyl-1,2-benzoquinone, 3,5-ditert-butylbenzo-1,2-quinone, 3,5-bis tert-butyl cyclohexa-3,5-diene-1,2-dione
PubChem CID 76915
IUPAC Name 3,5-ditert-butylcyclohexa-3,5-diene-1,2-dione
SMILES CC(C)(C)C1=CC(=O)C(=O)C(=C1)C(C)(C)C

Specifications

Melting Point 115°C
Quantity 5 g
Formula Weight 220.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Di-tert-butyl-1,2-benzoquinone