accessibility menu, dialog, popup

UserName

CAS RN 3383-21-9

IUPAC Name: 3,5-di-tert-butylcyclohexa-3,5-diene-1,2-dione
Synonyms: 3,5-di-tert-butylcyclohexa-3,5-diene-1,2-dione
Molecular Weight (g/mol): 220.31
Molecular Formula: C14H20O2
InChi Key: NOUZOVBGCDDMSX-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(=O)C(=O)C(=C1)C(C)(C)C
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)