missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Di-tert-butyl-1,2-benzoquinone 98.0+%, TCI America™
Supplier: TCI America D243025G
Specifications
| 3,5-Di-tert-butyl-1,2-benzoquinone | |
| 115°C | |
| MFCD00001647 | |
| 3,5-di-tert-butyl-o-benzoquinone, 3,5-di-tert-butyl-1,2-benzoquinone, 3,5-di tert-butyl benzo-1,2-quinone, 3,5-cyclohexadiene-1,2-dione, 3,5-bis 1,1-dimethylethyl, 3,5-di-tert-butylcyclohexa-3,5-diene-1,2-dione, o-benzoquinone, 3,5-di-tert-butyl, 3,5-di-tert-butylbenzo-1,2-quinone, 3,5-di-t-butyl-1,2-benzoquinone, 3,5-ditert-butylbenzo-1,2-quinone, 3,5-bis tert-butyl cyclohexa-3,5-diene-1,2-dione | |
| CC(C)(C)C1=CC(=O)C(=O)C(=C1)C(C)(C)C | |
| 220.312 | |
| 220.31 | |
| Crystalline Powder |
| 3383-21-9 | |
| C14H20O2 | |
| 25 g | |
| NOUZOVBGCDDMSX-UHFFFAOYSA-N | |
| 3,5-ditert-butylcyclohexa-3,5-diene-1,2-dione | |
| 76915 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 3383-21-9 | |
| 220.312 | |
| NOUZOVBGCDDMSX-UHFFFAOYSA-N | |
| 76915 | |
| CC(C)(C)C1=CC(=O)C(=O)C(=C1)C(C)(C)C |
| C14H20O2 | |
| MFCD00001647 | |
| 3,5-di-tert-butyl-o-benzoquinone, 3,5-di-tert-butyl-1,2-benzoquinone, 3,5-di tert-butyl benzo-1,2-quinone, 3,5-cyclohexadiene-1,2-dione, 3,5-bis 1,1-dimethylethyl, 3,5-di-tert-butylcyclohexa-3,5-diene-1,2-dione, o-benzoquinone, 3,5-di-tert-butyl, 3,5-di-tert-butylbenzo-1,2-quinone, 3,5-di-t-butyl-1,2-benzoquinone, 3,5-ditert-butylbenzo-1,2-quinone, 3,5-bis tert-butyl cyclohexa-3,5-diene-1,2-dione | |
| 3,5-ditert-butylcyclohexa-3,5-diene-1,2-dione |
Safety and Handling
TSCA : No