missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)thiophenol, 98%

CAS: 130783-02-7 | C8H4F6S | 246.17 g/mol

$568.42 - $568.42

Chemical Identifiers

CAS 130783-02-7
Molecular Formula C8H4F6S
Molecular Weight (g/mol) 246.17
MDL Number MFCD00042273
InChI Key KCAQWPZIMLLEAF-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl benzenethiol, 3,5-bis trifluoromethyl thiophenol, 3,5-bis trifluoromethyl benzene-1-thiol, pubchem15275, acmc-209bji, 3,5-bis trifluoromethyl-benzenethiol, 3,5-bis trifluoromethyl benzenethiol #
PubChem CID 518690
IUPAC Name 3,5-bis(trifluoromethyl)benzenethiol
SMILES C1=C(C=C(C=C1C(F)(F)F)S)C(F)(F)F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1466906
View Documents
Thermo Scientific Chemicals
A1466906
5 g
Each for $568.42
Only null left
 
Description

Description

3,5-Bis(trifluoromethyl)thiophenol, is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Bis(trifluoromethyl)thiophenol, is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Insoluble in water but soluble in alcohol and ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Air Sensitive.
Specifications

Chemical Identifiers

130783-02-7
246.17
KCAQWPZIMLLEAF-UHFFFAOYSA-N
518690
C1=C(C=C(C=C1C(F)(F)F)S)C(F)(F)F
C8H4F6S
MFCD00042273
3,5-bis trifluoromethyl benzenethiol, 3,5-bis trifluoromethyl thiophenol, 3,5-bis trifluoromethyl benzene-1-thiol, pubchem15275, acmc-209bji, 3,5-bis trifluoromethyl-benzenethiol, 3,5-bis trifluoromethyl benzenethiol #
3,5-bis(trifluoromethyl)benzenethiol

Specifications

130783-02-7
C8H4F6S
5 g
Air Sensitive
Insoluble in water but soluble in alcohol and ether.
C1=C(C=C(C=C1C(F)(F)F)S)C(F)(F)F
246.17
246.18
3,5-Bis(trifluoromethyl)thiophenol
63°C to 64°C (15 mmHg)
MFCD00042273
UN2810
3,5-bis trifluoromethyl benzenethiol, 3,5-bis trifluoromethyl thiophenol, 3,5-bis trifluoromethyl benzene-1-thiol, pubchem15275, acmc-209bji, 3,5-bis trifluoromethyl-benzenethiol, 3,5-bis trifluoromethyl benzenethiol #
KCAQWPZIMLLEAF-UHFFFAOYSA-N
3,5-bis(trifluoromethyl)benzenethiol
518690
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only