accessibility menu, dialog, popup

UserName

CAS RN 130783-02-7

IUPAC Name: [3,5-bis(trifluoromethyl)phenyl]sulfanide
Synonyms: [3,5-bis(trifluoromethyl)phenyl]sulfanide
Molecular Weight (g/mol): 245.16
Molecular Formula: C8H3F6S
InChi Key: KCAQWPZIMLLEAF-UHFFFAOYSA-M
SMILES: FC(F)(F)C1=CC(=CC([S-])=C1)C(F)(F)F
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)