missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)phenylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog No. p-7031786
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3297506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3297506 Supplier Thermo Scientific Chemicals Supplier No. H3297506

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 502496-23-3 | Molecular Formula: C8H7ClF6N2 | Molecular Weight: 280.598 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 502496-23-3
Molecular Formula C8H7ClF6N2
Molecular Weight (g/mol) 280.598
MDL Number MFCD03094165
InChI Key OPIOKSFIERNABH-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl phenylhydrazine hydrochloride, 3,5-ditrifluoromethylphenylhydrazine hydrochloride, 3,5-bis trifluoromethyl phenyl hydrazine hydrochloride, 3,5-bis trifluoromethyl phenylhydrazinehydrochloride, 3,5-bis trifluoromethy phenylhydrazine hcl, 3,5-bis trifluoromethyl phenylhydrazine hcl, ksc491e7p, 3,5-bis trifluoromethyl phenylhydrazine hydrochlor
PubChem CID 12494556
IUPAC Name [3,5-bis(trifluoromethyl)phenyl]hydrazine;hydrochloride
SMILES C1=C(C=C(C=C1C(F)(F)F)NN)C(F)(F)F.Cl

Specifications

Melting Point ∽208°C (decomposition)
Quantity 5 g
Formula Weight 280.6
Percent Purity 98%
Chemical Name or Material 3,5-Bis(trifluoromethyl)phenylhydrazine hydrochloride
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only