Learn More
3,5-Bis(trifluoromethyl)phenylacetonitrile, 97%
CAS: 85068-32-2 | C10H5F6N | 253.15 g/mol
$73.10 - $233.01
Chemical Identifiers
| CAS | 85068-32-2 |
|---|---|
| Molecular Formula | C10H5F6N |
| Molecular Weight (g/mol) | 253.15 |
| MDL Number | MFCD00009904 |
| InChI Key | YXGWYBUKRTYHJM-UHFFFAOYSA-N |
| Synonym | 3,5-bis trifluoromethyl phenylacetonitrile, 2-3,5-bis trifluoromethyl phenyl acetonitrile, 3,5-bis trifluoromethyl benzyl cyanide, 3,5-di trifluoromethyl benzyl cyanide, 3,5-bis trifluoromethyl-phenylacetonitrile, 3,5-bis trifluoromethyl phenyl acetonitrile, benzeneacetonitrile, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl benzylcyanide |
| PubChem CID | 603657 |
| IUPAC Name | 2-[3,5-bis(trifluoromethyl)phenyl]acetonitrile |
| SMILES | FC(F)(F)C1=CC(=CC(CC#N)=C1)C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1957603
|
Thermo Scientific Chemicals
A1957603 |
1 g |
Each for $73.10
|
|
|||||
|
AAA1957606
|
Thermo Scientific Chemicals
A1957606 |
5 g |
Each for $233.01
|
|
|||||
Description
3,5-Bis(trifluoromethyl)phenylacetonitrile may be used in chemical synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,5-Bis(trifluoromethyl)phenylacetonitrile may be used in chemical synthesis.
Solubility
Difficult to mix.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Chemical Identifiers
| 85068-32-2 | |
| 253.15 | |
| YXGWYBUKRTYHJM-UHFFFAOYSA-N | |
| 603657 | |
| FC(F)(F)C1=CC(=CC(CC#N)=C1)C(F)(F)F |
| C10H5F6N | |
| MFCD00009904 | |
| 3,5-bis trifluoromethyl phenylacetonitrile, 2-3,5-bis trifluoromethyl phenyl acetonitrile, 3,5-bis trifluoromethyl benzyl cyanide, 3,5-di trifluoromethyl benzyl cyanide, 3,5-bis trifluoromethyl-phenylacetonitrile, 3,5-bis trifluoromethyl phenyl acetonitrile, benzeneacetonitrile, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl benzylcyanide | |
| 2-[3,5-bis(trifluoromethyl)phenyl]acetonitrile |
Specifications
| 85068-32-2 | |
| >110°C (230°F) | |
| 1.4234 | |
| 1 g | |
| 6810362 | |
| Difficult to mix. | |
| FC(F)(F)C1=CC(=CC(CC#N)=C1)C(F)(F)F | |
| 253.15 | |
| 253.15 | |
| 3,5-Bis(trifluoromethyl)phenylacetonitrile |
| 1.42 | |
| C10H5F6N | |
| MFCD00009904 | |
| UN3276 | |
| 3,5-bis trifluoromethyl phenylacetonitrile, 2-3,5-bis trifluoromethyl phenyl acetonitrile, 3,5-bis trifluoromethyl benzyl cyanide, 3,5-di trifluoromethyl benzyl cyanide, 3,5-bis trifluoromethyl-phenylacetonitrile, 3,5-bis trifluoromethyl phenyl acetonitrile, benzeneacetonitrile, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl benzylcyanide | |
| YXGWYBUKRTYHJM-UHFFFAOYSA-N | |
| 2-[3,5-bis(trifluoromethyl)phenyl]acetonitrile | |
| 603657 | |
| 97% |
Safety and Handling
GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
EINECSNumber : 285-294-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only