missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)phenylacetonitrile, 97%

Catalog No. AAA1957603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1957603 1 g
AAA1957606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1957603 Supplier Thermo Scientific Chemicals Supplier No. A1957603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 85068-32-2 | C10H5F6N | 253.15 g/mol

3,5-Bis(trifluoromethyl)phenylacetonitrile may be used in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Bis(trifluoromethyl)phenylacetonitrile may be used in chemical synthesis.

Solubility
Difficult to mix.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 85068-32-2
Molecular Formula C10H5F6N
Molecular Weight (g/mol) 253.15
MDL Number MFCD00009904
InChI Key YXGWYBUKRTYHJM-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl phenylacetonitrile, 2-3,5-bis trifluoromethyl phenyl acetonitrile, 3,5-bis trifluoromethyl benzyl cyanide, 3,5-di trifluoromethyl benzyl cyanide, 3,5-bis trifluoromethyl-phenylacetonitrile, 3,5-bis trifluoromethyl phenyl acetonitrile, benzeneacetonitrile, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl benzylcyanide
PubChem CID 603657
IUPAC Name 2-[3,5-bis(trifluoromethyl)phenyl]acetonitrile
SMILES FC(F)(F)C1=CC(=CC(CC#N)=C1)C(F)(F)F

Specifications

Density 1.42
Flash Point >110°C (230°F)
Refractive Index 1.4234
Quantity 1 g
UN Number UN3276
Beilstein 6810362
Solubility Information Difficult to mix.
Formula Weight 253.15
Percent Purity 97%
Chemical Name or Material 3,5-Bis(trifluoromethyl)phenylacetonitrile
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
EINECSNumber 285-294-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only