missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)-o-phenylenediamine, 97%

Catalog No. AAL1818006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1818006 5 g
AAL1818003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1818006 Supplier Thermo Scientific Chemicals Supplier No. L1818006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 367-65-7 | C8H6F6N2 | 244.14 g/mol

It is applied as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied as an active pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 367-65-7
Molecular Formula C8H6F6N2
Molecular Weight (g/mol) 244.14
MDL Number MFCD01631430
InChI Key BRLIJPMFMGTIAW-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl-1,2-diaminobenzene, 3,5-bis trifluoromethyl benzene-1,2-diamine, 3,5-bis trifluoromethyl-1,2-phenylenediamine, 1,2-diamino-3,5-bis trifluoromethyl benzene, 1,2-benzenediamine, 3,5-bis trifluoromethyl, acmc-1cr3d, 3,5-bis trifluoromethyl-o-phenylenediamine, 1,2-benzenediamine,3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl phenylene-1,2-diamine
PubChem CID 2773214
IUPAC Name 3,5-bis(trifluoromethyl)benzene-1,2-diamine
SMILES NC1=CC(=CC(=C1N)C(F)(F)F)C(F)(F)F

Specifications

Melting Point 44°C to 49°C
Quantity 5 g
UN Number UN2811
Beilstein 3344332
Solubility Information Insoluble in water.
Formula Weight 244.14
Percent Purity 97%
Chemical Name or Material 3,5-Bis(trifluoromethyl)-o-phenylenediamine
Hazard Category H302+H332-H311-H315-H319-H335
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only