accessibility menu, dialog, popup

UserName

CAS RN 367-65-7

IUPAC Name: 3,5-bis(trifluoromethyl)benzene-1,2-diamine
Synonyms: 3,5-bis(trifluoromethyl)benzene-1,2-diamine
Molecular Weight (g/mol): 244.14
Molecular Formula: C8H6F6N2
InChi Key: BRLIJPMFMGTIAW-UHFFFAOYSA-N
SMILES: NC1=CC(=CC(=C1N)C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results