missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)bromobenzene, 99%

Catalog No. AC307702500
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC307702500 250 g Glass bottle
AC307700100 10 g Glass bottle
AC307700500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC307702500 Supplier Thermo Scientific Chemicals Supplier No. 307702500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 328-70-1 | C8H3BrF6 | 293.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 328-70-1
Molecular Formula C8H3BrF6
Molecular Weight (g/mol) 293.01
InChI Key CSVCVIHEBDJTCJ-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl bromobenzene, 1-bromo-3,5-bis trifluoromethyl benzene, 1,3-bis trifluoromethyl-5-bromobenzene, mbt-br, 3,5-di trifluoromethyl bromobenzene, 3,5-bis trifluoromethyl-1-bromobenzene, 3,5-bis-trifluoromethylbromobenzene, benzene, 1-bromo-3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl phenyl bromide, 1-bromo-3,5-bis-trifluoromethyl-benzene
PubChem CID 67602
IUPAC Name 1-bromo-3,5-bis(trifluoromethyl)benzene
SMILES C1=C(C=C(C=C1C(F)(F)F)Br)C(F)(F)F

Specifications

Melting Point -16.0°C
Color Colorless to Yellow
Density 1.7100g/mL
Boiling Point 154.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4260 to 1.4280
Linear Formula (CF3)2C6H3Br
Quantity 250 g
Specific Gravity 1.71
Formula Weight 293.01
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3, 5-Bis(trifluoromethyl)bromobenzene
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 206-334-5

RUO – Research Use Only