accessibility menu, dialog, popup

UserName

FFFBrFFF

CAS RN 328-70-1

IUPAC Name: 1-bromo-3,5-bis(trifluoromethyl)benzene
Synonyms: 1-bromo-3,5-bis(trifluoromethyl)benzene
Molecular Weight (g/mol): 293.01
Molecular Formula: C8H3BrF6
InChi Key: CSVCVIHEBDJTCJ-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(=CC(Br)=C1)C(F)(F)F
Boiling Point 
  • (2)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (3)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (2)
  • (3)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (5)