missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)benzyl Chloride 97.0+%, TCI America™

Catalog No. B36415G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36415G 5 g
B364125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36415G Supplier TCI America Supplier No. B36415G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 75462-59-8
Molecular Formula C9H5ClF6
Molecular Weight (g/mol) 262.579
MDL Number MFCD00009906
InChI Key OINTXXMBRBLMHH-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl benzyl chloride, 1-chloromethyl-3,5-bis trifluoromethyl benzene, 3,5-bis-trifluoromethyl benzyl chloride, 3,5-bis trifluoromethyl benzylchloride, 3,5-di trifluoromethyl benzyl chloride, benzene, 1-chloromethyl-3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl-1-chloromethyl benzene, 1,3-bis trifluoromethyl-5-chloromethyl benzene
PubChem CID 123576
IUPAC Name 1-(chloromethyl)-3,5-bis(trifluoromethyl)benzene
SMILES C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)CCl

Specifications

Color Yellow
Boiling Point 84°C
Quantity 5 g
UN Number 3265
Formula Weight 262.58
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,5-Bis(trifluoromethyl)benzyl Chloride