missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)benzyl bromide, 97%

Catalog No. AC303640050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC303640050 5 g Glass bottle
AC303640010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC303640050 Supplier Thermo Scientific Chemicals Supplier No. 303640050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 32247-96-4 | C9H5BrF6 | 307.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 32247-96-4
Molecular Formula C9H5BrF6
Molecular Weight (g/mol) 307.03
MDL Number MFCD00009905
InChI Key ATLQGZVLWOURFU-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl benzyl bromide, 1-bromomethyl-3,5-bis trifluoromethyl benzene, 3,5-di trifluoromethyl benzyl bromide, 3,5-bis trifluoromethyl benzylbromide, 1-bromomethyl-3,5-di trifluoromethyl benzene, 3,5-bis-trifluoromethyl benzyl bromide, benzene, 1-bromomethyl-3,5-bis trifluoromethyl, 1,3-bis trifluoromethyl-5-bromomethyl benzene
PubChem CID 122573
IUPAC Name 1-(bromomethyl)-3,5-bis(trifluoromethyl)benzene
SMILES FC(F)(F)C1=CC(=CC(CBr)=C1)C(F)(F)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Density 1.6700g/mL
Flash Point 26°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CF3)2C6H3CH2Br
Refractive Index 1.4440 to 1.446
Quantity 5 g
Specific Gravity 1.67
Formula Weight 307.04
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3, 5-Bis(trifluoromethyl)benzyl bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 250-971-1

RUO – Research Use Only