accessibility menu, dialog, popup

UserName

FFFBrFFF

CAS RN 32247-96-4

IUPAC Name: 1-(bromomethyl)-3,5-bis(trifluoromethyl)benzene
Synonyms: 1-(bromomethyl)-3,5-bis(trifluoromethyl)benzene
Molecular Weight (g/mol): 307.03
Molecular Formula: C9H5BrF6
InChi Key: ATLQGZVLWOURFU-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(=CC(CBr)=C1)C(F)(F)F
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)