missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)benzenethiol 96.0+%, TCI America™

$136.08 - $447.38

Chemical Identifiers

CAS 130783-02-7
Molecular Formula C8H4F6S
Molecular Weight (g/mol) 246.17
MDL Number MFCD00042273
InChI Key KCAQWPZIMLLEAF-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl benzenethiol, 3,5-bis trifluoromethyl thiophenol, 3,5-bis trifluoromethyl benzene-1-thiol, pubchem15275, acmc-209bji, 3,5-bis trifluoromethyl-benzenethiol, 3,5-bis trifluoromethyl benzenethiol #
PubChem CID 518690
IUPAC Name 3,5-bis(trifluoromethyl)benzenethiol
SMILES C1=C(C=C(C=C1C(F)(F)F)S)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B23911G
View Documents
TCI America
B23911G
1 g
Each for $136.08
Only null left
 
B23915G
View Documents
TCI America
B23915G
5 g
Each for $447.38
Only null left
 
Description

Description

Specifications

Chemical Identifiers

130783-02-7
246.17
KCAQWPZIMLLEAF-UHFFFAOYSA-N
518690
C1=C(C=C(C=C1C(F)(F)F)S)C(F)(F)F
C8H4F6S
MFCD00042273
3,5-bis trifluoromethyl benzenethiol, 3,5-bis trifluoromethyl thiophenol, 3,5-bis trifluoromethyl benzene-1-thiol, pubchem15275, acmc-209bji, 3,5-bis trifluoromethyl-benzenethiol, 3,5-bis trifluoromethyl benzenethiol #
3,5-bis(trifluoromethyl)benzenethiol

Specifications

130783-02-7
167°C
MFCD00042273
3,5-bis trifluoromethyl benzenethiol, 3,5-bis trifluoromethyl thiophenol, 3,5-bis trifluoromethyl benzene-1-thiol, pubchem15275, acmc-209bji, 3,5-bis trifluoromethyl-benzenethiol, 3,5-bis trifluoromethyl benzenethiol #
C1=C(C=C(C=C1C(F)(F)F)S)C(F)(F)F
246.17
246.17
Liquid
Yellow
C8H4F6S
1 g
KCAQWPZIMLLEAF-UHFFFAOYSA-N
3,5-bis(trifluoromethyl)benzenethiol
518690
≥96.0% (GC,T)
3,5-Bis(trifluoromethyl)benzenethiol
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'