missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)benzenethiol 96.0+%, TCI America™

Catalog No. B23911G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23911G 1 g
B23915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B23911G Supplier TCI America Supplier No. B23911G

Chemical Identifiers

CAS 130783-02-7
Molecular Formula C8H4F6S
Molecular Weight (g/mol) 246.17
MDL Number MFCD00042273
InChI Key KCAQWPZIMLLEAF-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl benzenethiol, 3,5-bis trifluoromethyl thiophenol, 3,5-bis trifluoromethyl benzene-1-thiol, pubchem15275, acmc-209bji, 3,5-bis trifluoromethyl-benzenethiol, 3,5-bis trifluoromethyl benzenethiol #
PubChem CID 518690
IUPAC Name 3,5-bis(trifluoromethyl)benzenethiol
SMILES C1=C(C=C(C=C1C(F)(F)F)S)C(F)(F)F

Specifications

Color Yellow
Boiling Point 167°C
Quantity 1 g
Formula Weight 246.17
Percent Purity ≥96.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3,5-Bis(trifluoromethyl)benzenethiol