missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(tert-butylthio)-1-chlorobenzene 98.0+%, TCI America™

Catalog No. B26591G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B26591G 1 g
B26595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B26591G Supplier TCI America Supplier No. B26591G
Only null left

Chemical Identifiers

CAS 260968-02-3
Molecular Formula C14H21ClS2
Molecular Weight (g/mol) 288.892
InChI Key QVEUHOXOWGXTJG-UHFFFAOYSA-N
Synonym 1-Chloro-3,5-bis(tert-butylthio)benzene
PubChem CID 22617204
IUPAC Name 1,3-bis(tert-butylsulfanyl)-5-chlorobenzene
SMILES CC(C)(C)SC1=CC(=CC(=C1)Cl)SC(C)(C)C

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 288.89
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Bis(tert-butylthio)-1-chlorobenzene