accessibility menu, dialog, popup

UserName

H3CH3CCH3SClSCH3CH3H3C

CAS RN 260968-02-3

IUPAC Name: 1,3-bis(tert-butylsulfanyl)-5-chlorobenzene
Synonyms: 1,3-bis(tert-butylsulfanyl)-5-chlorobenzene
Molecular Weight (g/mol): 288.89
Molecular Formula: C14H21ClS2
InChi Key: QVEUHOXOWGXTJG-UHFFFAOYSA-N
SMILES: CC(C)(C)SC1=CC(SC(C)(C)C)=CC(Cl)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results