missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(3,5-dimethoxybenzyloxy)benzyl Bromide 97.0+%, TCI America™

Catalog No. B22475G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22475G 5 g
B22471G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B22475G Supplier TCI America Supplier No. B22475G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 176650-93-4
Molecular Formula C25H27BrO6
Molecular Weight (g/mol) 503.39
MDL Number MFCD04038410
InChI Key RLACLNXCSSBIKS-UHFFFAOYSA-N
PubChem CID 15778113
IUPAC Name 1-{[3-(bromomethyl)-5-[(3,5-dimethoxyphenyl)methoxy]phenoxy]methyl}-3,5-dimethoxybenzene
SMILES COC1=CC(OC)=CC(COC2=CC(OCC3=CC(OC)=CC(OC)=C3)=CC(CBr)=C2)=C1

Specifications

Quantity 5 g
Formula Weight 503.39
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Bis(3,5-dimethoxybenzyloxy)benzyl Bromide
TSCA No
Recommended Storage Refrigerator